siRNA Details
| Gene Symbol: LOC100911147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748752.1 | XP_003748800.1 | 3 | 1099 | ||
| XM_006228209.1 | XP_006228271.1 | 3 | 1099 | |||
| XM_006228210.1 | XP_006228272.1 | 3 | 1099 | |||
| XM_006228211.1 | XP_006228273.1 | 3 | 1099 | |||
| Gene Symbol: Vom2r71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099516.1 | NP_001092986.1 | 1105 | |||
More Information
Set Membership: |
|

LOC100911147
Genomic Map