siRNA Details
| Gene Symbol: C1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106021.1 | NP_001099491.1 | 3 | 287 | ||
| XM_006251516.2 | XP_006251578.1 | 4 | 406 | |||
| XM_008770422.1 | XP_008768644.1 | 3 | 2469 | |||
| XM_008770423.1 | XP_008768645.1 | 4 | 2368 | |||
| GenBank mRNA | BC168684.1 | - | 3 | 263 | 7930158 | |
| FQ215730.1 | - | 3 | 261 | |||
| FQ229879.1 | - | 3 | 216 | |||
| FQ229896.1 | - | 3 | 265 | |||

C1d
Genomic Map