siRNA Details
| Gene Symbol: MRC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006039.4 | NP_006030.2 | 22 | 3488 | ||
| XM_011525543.2 | XP_011523845.1 | 22 | 3219 | |||
| GenBank mRNA | AB014609.2 | - | 22 | 3488 | ||
| AF107292.1 | - | 22 | 3129 | |||
| AF134838.1 | - | 22 | 3210 | |||
| AK093230.1 | - | 4 | 942 | |||
| BC146647.1 | - | 22 | 3207 | 40147137 | ||
| BC150212.1 | - | 22 | 3192 | 40148946 | ||
| BC153884.1 | - | 22 | 3488 | 8860500 | ||

MRC2
Genomic Map