siRNA Details
| Gene Symbol: Grm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105712.1 | NP_001099182.1 | 2 | 200 | ||
| XM_008762681.1 | XP_008760903.1 | 1 | 1893 | |||
| XM_008762682.1 | XP_008760904.1 | 2 | 676 | |||
| XM_008762683.1 | XP_008760905.1 | 1 | 1893 | |||
| GenBank mRNA | M92076.1 | - | 1 | 118 | ||
More Information
Set Membership: |
|

Grm3
Genomic Map