siRNA Details
| Gene Symbol: KLHL21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001324309.1 | NP_001311238.1 | 4 | 2102 | ||
| NM_014851.3 | NP_055666.2 | 4 | 1650 | |||
| GenBank mRNA | AB007938.1 | - | 3 | 3579 | ||
| AK075305.1 | - | 4 | 1621 | |||
| AK090472.1 | - | 4 | 1419 | |||
| AK307564.1 | - | 4 | 610 | |||
| BC034039.1 | - | 4 | 1621 | 4811534 | ||
| BC064980.1 | - | 4 | 1603 | |||
| BC091648.1 | - | 4 | 1603 | 6373903 | ||
| BC111766.1 | - | 2 | 1559 | |||

KLHL21
Genomic Map