siRNA Details
| Gene Symbol: Snx29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109526.2 | NP_001102996.2 | 21 | 2390 | ||
| XM_006245736.2 | XP_006245798.1 | 20 | 2696 | |||
| XM_008767497.1 | XP_008765719.1 | 21 | 3505 | |||
| XM_008767498.1 | XP_008765720.1 | 19 | 2331 | |||
| XM_008767499.1 | XP_008765721.1 | 14 | 3908 | |||

Snx29
Genomic Map