siRNA Details
Gene Symbol: LOC690806 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135576.2 | NP_001129048.1 | 218 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC690806 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135576.2 | NP_001129048.1 | 218 |