siRNA Details
| Gene Symbol: Ccdc87 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109527.2 | NP_001102997.1 | 1 | 1792 | ||
| GenBank mRNA | BC166526.1 | - | 1 | 1792 | 7135699 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc87| Gene Symbol: Ccdc87 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109527.2 | NP_001102997.1 | 1 | 1792 | ||
| GenBank mRNA | BC166526.1 | - | 1 | 1792 | 7135699 | |