siRNA Details
| Gene Symbol: AP5Z1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014855.2 | NP_055670.1 | 9 | 1201 | ||
| XM_017012864.1 | XP_016868353.1 | 8 | 1028 | |||
| GenBank mRNA | AB007875.1 | - | 9 | 1180 | ||
| BC037399.1 | - | 7 | 874 | 5172434 | ||
| BQ228738.1 | - | 227 | ||||

AP5Z1
Genomic Map