siRNA Details
| Gene Symbol: Mis18bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109531.1 | NP_001103001.1 | 5, 6 | 1488 | ||
| XM_006240138.2 | XP_006240200.1 | 5, 6 | 2393 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mis18bp1| Gene Symbol: Mis18bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109531.1 | NP_001103001.1 | 5, 6 | 1488 | ||
| XM_006240138.2 | XP_006240200.1 | 5, 6 | 2393 | |||