siRNA Details
| Gene Symbol: Fam159a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001070397.5 | XP_001070397.2 | 2 | 479 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam159a| Gene Symbol: Fam159a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001070397.5 | XP_001070397.2 | 2 | 479 | ||