siRNA Details
| Gene Symbol: Loh12cr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006237528.2 | XP_006237590.1 | 1 | 541 | ||
| XM_008763394.1 | XP_008761616.1 | 1 | 539 | |||
| XM_008763395.1 | XP_008761617.1 | 1 | 539 | |||
| XM_008763396.1 | XP_008761618.1 | 1 | 541 | |||

Loh12cr1
Genomic Map