siRNA Details
Gene Symbol: Mat2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044282.1 | NP_001037747.1 | 4 | 557 | ||
XM_006246126.2 | XP_006246188.1 | 4 | 707 | |||
GenBank mRNA | BC085899.1 | - | 4 | 557 | 7193416 |
