siRNA Details
| Gene Symbol: Ptx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109536.1 | NP_001103006.1 | 3 | 1176 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptx3| Gene Symbol: Ptx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109536.1 | NP_001103006.1 | 3 | 1176 | ||