siRNA Details
| Gene Symbol: Dram2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025018.1 | NP_001020189.1 | 7 | 749 | ||
| XM_006233120.2 | XP_006233182.1 | 7 | 693 | |||
| XM_006233121.2 | XP_006233183.1 | 6 | 633 | |||
| XM_006233122.2 | XP_006233184.1 | 6 | 734 | |||
| XM_006233123.2 | XP_006233185.1 | 7 | 695 | |||
| GenBank mRNA | BC091251.1 | - | 7 | 749 | 7372838 | |
| FQ218349.1 | - | 7 | 758 | |||
| FQ228782.1 | - | 7 | 846 | |||
| Gene Symbol: LOC689412 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109538.1 | NP_001103008.1 | 1 | 396 | ||

Dram2
Genomic Map