siRNA Details
| Gene Symbol: Cnbd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763603.1 | XP_008761825.1 | 4 | 574 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnbd1| Gene Symbol: Cnbd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763603.1 | XP_008761825.1 | 4 | 574 | ||