siRNA Details
| Gene Symbol: Calr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763952.1 | XP_008762174.1 | 3 | 535 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Calr4| Gene Symbol: Calr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763952.1 | XP_008762174.1 | 3 | 535 | ||