siRNA Details
Gene Symbol: Calr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763952.1 | XP_008762174.1 | 4 | 617 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
