siRNA Details
| Gene Symbol: FAM20B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001324310.1 | NP_001311239.1 | 5 | 847 | ||
| NM_001324311.1 | NP_001311240.1 | 5 | 857 | |||
| NM_014864.3 | NP_055679.1 | 5 | 952 | |||
| XM_017003001.1 | XP_016858490.1 | 6 | 898 | |||
| XM_017003002.1 | XP_016858491.1 | 6 | 891 | |||
| GenBank mRNA | AB007944.1 | - | 5 | 995 | ||
| AB480690.1 | - | 5 | 952 | |||
| AK289989.1 | - | 5 | 852 | |||
| BC046441.1 | - | 5 | 820 | 5197154 | ||
| BC051794.1 | - | 5 | 952 | 5492043 | ||

FAM20B
Genomic Map