siRNA Details
| Gene Symbol: Psmg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129879.2 | NP_001123351.1 | 1, 2 | 274 | ||
| XM_006253847.2 | XP_006253909.1 | 1, 2 | 282 | |||
| GenBank mRNA | BC167096.1 | - | 1, 2 | 201 | 8362489 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psmg4| Gene Symbol: Psmg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129879.2 | NP_001123351.1 | 1, 2 | 274 | ||
| XM_006253847.2 | XP_006253909.1 | 1, 2 | 282 | |||
| GenBank mRNA | BC167096.1 | - | 1, 2 | 201 | 8362489 | |