siRNA Details
| Gene Symbol: Vom2r70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099477.1 | NP_001092947.1 | 6 | 2231 | ||
| Gene Symbol: Vom2r73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099486.1 | NP_001092956.1 | 6 | 2231 | ||
More Information
Set Membership: |
|

Vom2r70
Genomic Map