siRNA Details
| Gene Symbol: Vom2r73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099486.1 | NP_001092956.1 | 3 | 533 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vom2r73| Gene Symbol: Vom2r73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099486.1 | NP_001092956.1 | 3 | 533 | ||