siRNA Details
| Gene Symbol: LOC689713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047978.1 | NP_001041443.1 | 2 | 86 | ||
| GenBank mRNA | AY539926.1 | - | 2 | 86 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC689713| Gene Symbol: LOC689713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047978.1 | NP_001041443.1 | 2 | 86 | ||
| GenBank mRNA | AY539926.1 | - | 2 | 86 | ||