siRNA Details
| Gene Symbol: Klrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085405.1 | NP_001078874.1 | 2, 3 | 175 | ||
| XM_008763334.1 | XP_008761556.1 | 2, 3 | 233 | |||
| XM_008763335.1 | XP_008761557.1 | 2, 3 | 233 | |||
| GenBank mRNA | DQ113420.1 | - | 2, 3 | 175 | ||
| DQ157011.1 | - | 2, 3 | 175 | |||
| DQ157012.1 | - | 2, 3 | 175 | |||
| EF100680.1 | - | 2, 3 | 175 | |||

Klrb1
Genomic Map