siRNA Details
| Gene Symbol: LOC689840 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047979.1 | NP_001041444.1 | 1 | 191 | ||
| GenBank mRNA | AY539893.1 | - | 1 | 191 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC689840| Gene Symbol: LOC689840 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047979.1 | NP_001041444.1 | 1 | 191 | ||
| GenBank mRNA | AY539893.1 | - | 1 | 191 | ||