siRNA Details
| Gene Symbol: ZBTB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014872.2 | NP_055687.1 | 2 | 1423 | ||
| XM_005251634.1 | XP_005251691.1 | 2 | 2267 | |||
| GenBank mRNA | AB002352.1 | - | 2 | 1412 | ||
| AK292404.1 | - | 2 | 1468 | |||
| BC010007.2 | - | 2 | 1267 | 4300173 | ||

ZBTB5
Genomic Map