siRNA Details
| Gene Symbol: Ankrd61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044297.1 | NP_001037762.1 | 3 | 946 | ||
| GenBank mRNA | BC082099.1 | - | 3 | 946 | 7134466 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankrd61| Gene Symbol: Ankrd61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044297.1 | NP_001037762.1 | 3 | 946 | ||
| GenBank mRNA | BC082099.1 | - | 3 | 946 | 7134466 | |