siRNA Details
Gene Symbol: Ccdc62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134766.1 | NP_001128238.1 | 9 | 1546 | ||
XM_006249355.2 | XP_006249417.1 | 9 | 2414 | |||
XM_006249356.2 | XP_006249418.1 | 9 | 2360 | |||
XM_006249357.2 | XP_006249419.1 | 9 | 2414 | |||
XM_006249358.1 | XP_006249420.1 | 9 | 1487 | |||
XM_008769265.1 | XP_008767487.1 | 8 | 1299 | |||
GenBank mRNA | BC086549.1 | - | 5 | 723 | ||
BC099139.1 | - | 5 | 825 |
