siRNA Details
Gene Symbol: Gpr81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145334.1 | NP_001138806.1 | 1 | 178 | ||
GenBank mRNA | EU809461.1 | - | 1 | 178 | ||
EU809462.1 | - | 1 | 154 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gpr81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145334.1 | NP_001138806.1 | 1 | 178 | ||
GenBank mRNA | EU809461.1 | - | 1 | 178 | ||
EU809462.1 | - | 1 | 154 |