siRNA Details
| Gene Symbol: Ndufc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749275.2 | XP_003749323.2 | 2 | 305 | ||
| XM_008761014.1 | XP_008759236.1 | 2 | 305 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ndufc1| Gene Symbol: Ndufc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749275.2 | XP_003749323.2 | 2 | 305 | ||
| XM_008761014.1 | XP_008759236.1 | 2 | 305 | |||