siRNA Details
| Gene Symbol: Gfi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769919.1 | XP_008768141.1 | 1 | 250 | ||
| XM_008769920.1 | XP_008768142.1 | 1 | 250 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gfi1| Gene Symbol: Gfi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769919.1 | XP_008768141.1 | 1 | 250 | ||
| XM_008769920.1 | XP_008768142.1 | 1 | 250 | |||