siRNA Details
| Gene Symbol: Nrsn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109561.1 | NP_001103031.1 | 3 | 366 | ||
| GenBank mRNA | BC159407.1 | - | 3 | 347 | 8362039 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nrsn2| Gene Symbol: Nrsn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109561.1 | NP_001103031.1 | 3 | 366 | ||
| GenBank mRNA | BC159407.1 | - | 3 | 347 | 8362039 | |