siRNA Details
| Gene Symbol: LOC689986 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109563.1 | NP_001103033.1 | 3, 4 | 2455 | ||
| XM_006250572.2 | XP_006250634.1 | 4, 5 | 722 | |||
| XM_006250573.2 | XP_006250635.1 | 4, 5 | 726 | |||
| XM_006250574.2 | XP_006250636.1 | 4, 5 | 626 | |||
| XM_006250575.2 | XP_006250637.1 | 3, 4 | 573 | |||
| XM_006250576.2 | XP_006250638.1 | 3, 4 | 477 | |||
| XM_006250580.2 | XP_006250642.1 | 4, 5 | 721 | |||
| XM_006250581.2 | XP_006250643.1 | 4, 5 | 723 | |||
| XM_008769942.1 | XP_008768164.1 | 5, 6 | 770 | |||

LOC689986
Genomic Map