siRNA Details
| Gene Symbol: LOC689986 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109563.1 | NP_001103033.1 | 1 | 801 | ||
| Gene Symbol: LOC100364240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006250596.2 | XP_006250658.2 | 17 | 3520 | ||
| XM_006250597.2 | XP_006250659.2 | 17 | 3517 | |||

LOC689986
Genomic Map