siRNA Details
| Gene Symbol: LOC690140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761828.1 | XP_008760050.1 | 2 | 606 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC690140| Gene Symbol: LOC690140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761828.1 | XP_008760050.1 | 2 | 606 | ||