siRNA Details
| Gene Symbol: Prima1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108721.2 | NP_001102191.2 | 5 | 1023 | ||
| XM_006240495.2 | XP_006240557.1 | 5 | 1466 | |||
| XM_008764849.1 | XP_008763071.1 | 5 | 1096 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prima1| Gene Symbol: Prima1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108721.2 | NP_001102191.2 | 5 | 1023 | ||
| XM_006240495.2 | XP_006240557.1 | 5 | 1466 | |||
| XM_008764849.1 | XP_008763071.1 | 5 | 1096 | |||