siRNA Details
| Gene Symbol: Slc16a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109568.1 | NP_001103038.1 | 3 | 798 | ||
| XM_006247735.2 | XP_006247797.1 | 3 | 612 | |||
| XM_008768396.1 | XP_008766618.1 | 3 | 772 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc16a5| Gene Symbol: Slc16a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109568.1 | NP_001103038.1 | 3 | 798 | ||
| XM_006247735.2 | XP_006247797.1 | 3 | 612 | |||
| XM_008768396.1 | XP_008766618.1 | 3 | 772 | |||