siRNA Details
| Gene Symbol: Wdr74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006230999.2 | XP_006231061.1 | 9, 10 | 960 | ||
| XM_006231000.2 | XP_006231062.1 | 9, 10 | 828 | |||
| GenBank mRNA | BC158834.1 | - | 9, 10 | 922 | 7383724 | |

Wdr74
Genomic Map