siRNA Details
| Gene Symbol: Usp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166576.1 | NP_001160048.1 | 7 | 803 | ||
| GenBank mRNA | FN557299.1 | - | 7 | 803 | ||
| Gene Symbol: LOC100912059 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751125.2 | XP_003751173.1 | 2 | 791 | ||
| Gene Symbol: RGD1561481 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_577055.5 | XP_577055.4 | 2 | 791 | ||

Usp12
Genomic Map