siRNA Details
| Gene Symbol: Magohb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191105.1 | NP_001178034.1 | 3 | 200 | ||
| XM_008763423.1 | XP_008761645.1 | 3 | 1476 | |||
| XM_008763424.1 | XP_008761646.1 | 4 | 345 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Magohb| Gene Symbol: Magohb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191105.1 | NP_001178034.1 | 3 | 200 | ||
| XM_008763423.1 | XP_008761645.1 | 3 | 1476 | |||
| XM_008763424.1 | XP_008761646.1 | 4 | 345 | |||