siRNA Details
| Gene Symbol: Sertm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109580.1 | NP_001103050.1 | 3 | 590 | ||
| XM_006232369.2 | XP_006232431.1 | 4 | 1012 | |||
| XM_006232370.2 | XP_006232432.1 | 5 | 1137 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sertm1| Gene Symbol: Sertm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109580.1 | NP_001103050.1 | 3 | 590 | ||
| XM_006232369.2 | XP_006232431.1 | 4 | 1012 | |||
| XM_006232370.2 | XP_006232432.1 | 5 | 1137 | |||