siRNA Details
| Gene Symbol: Tex12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766337.1 | XP_008764559.1 | 1 | 2798 | ||
| XM_008766338.1 | XP_008764560.1 | 1 | 2798 | |||
| XM_008766339.1 | XP_008764561.1 | 2 | 2434 | |||
| Gene Symbol: Bco2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY325176.1 | - | 99 | |||

Tex12
Genomic Map