siRNA Details
Gene Symbol: Rbbp8nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762592.1 | XP_008760814.1 | 6, 7 | 1985 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rbbp8nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762592.1 | XP_008760814.1 | 6, 7 | 1985 |