siRNA Details
| Gene Symbol: LOC100909526 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765141.1 | XP_008763363.1 | 3 | 2237 | ||
| Gene Symbol: Zfp709l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163063.1 | NP_001156535.1 | 4 | 1286 | ||

LOC100909526
Genomic Map