siRNA Details
| Gene Symbol: OXSR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005109.2 | NP_005100.1 | 17 | 1824 | ||
| XM_005265638.2 | XP_005265695.1 | 18 | 1965 | |||
| XM_011534331.2 | XP_011532633.1 | 17 | 1876 | |||
| XM_017007601.1 | XP_016863090.1 | 16 | 1631 | |||
| XM_017007602.1 | XP_016863091.1 | 19 | 2151 | |||
| GenBank mRNA | AB017642.1 | - | 17 | 1824 | ||
| AB029024.1 | - | 17 | 1577 | |||
| BC008726.2 | - | 17 | 1807 | 3163379 | ||

OXSR1
Genomic Map