siRNA Details
| Gene Symbol: Tmem116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159625.1 | NP_001153097.1 | 5 | 300 | ||
| XM_006249404.2 | XP_006249466.1 | 5 | 650 | |||
| GenBank mRNA | BC167104.1 | - | 5 | 300 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem116| Gene Symbol: Tmem116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159625.1 | NP_001153097.1 | 5 | 300 | ||
| XM_006249404.2 | XP_006249466.1 | 5 | 650 | |||
| GenBank mRNA | BC167104.1 | - | 5 | 300 | ||