siRNA Details
| Gene Symbol: Vom2r12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099488.1 | NP_001092958.1 | 3 | 515 | ||
| Gene Symbol: LOC100910184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768625.1 | XP_008766847.1 | 5 | 683 | ||
More Information
Set Membership: |
|

Vom2r12
Genomic Map