siRNA Details
Gene Symbol: Tcl1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109601.1 | NP_001103071.1 | 2 | 338 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tcl1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109601.1 | NP_001103071.1 | 2 | 338 |