siRNA Details
| Gene Symbol: Alyref | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109602.1 | NP_001103072.1 | 3 | 484 | ||
| XM_006247925.2 | XP_006247987.1 | 3 | 479 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alyref| Gene Symbol: Alyref | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109602.1 | NP_001103072.1 | 3 | 484 | ||
| XM_006247925.2 | XP_006247987.1 | 3 | 479 | |||