siRNA Details
| Gene Symbol: Fbxo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109606.1 | NP_001103076.1 | 4 | 461 | ||
| XM_006234656.2 | XP_006234718.1 | 4 | 486 | |||
| XM_006234657.2 | XP_006234719.1 | 4 | 486 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxo3| Gene Symbol: Fbxo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109606.1 | NP_001103076.1 | 4 | 461 | ||
| XM_006234656.2 | XP_006234718.1 | 4 | 486 | |||
| XM_006234657.2 | XP_006234719.1 | 4 | 486 | |||